missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Glutamic Acid, free acid, ≥99%, MP Biomedicals™
Supplier: MP Biomedicals Inc 0210180080
Specifications
L-Glutamic acid Monosodium Salt | |
White | |
C5H8NNaO4 | |
natriumglutaminat, sodium glutamate, glutamate sodium, monosodioglutammato, sodium l-glutamate, glutammato monosodico, monosodium l-glutamate, sodium hydrogen glutamate, monosodium glutamate, glutamic acid, monosodium salt | |
[Na+].NC(CCC(O)=O)C([O-])=O | |
169.11 | |
187.13 | |
100 g |
142-47-2 | |
≥99% | |
MFCD00150138 | |
LPUQAYUQRXPFSQ-UHFFFAOYNA-M | |
sodium 2-amino-4-carboxybutanoate | |
86748263 | |
1.538g/mL at 20°C | |
Powder |
Chemical Identifiers
142-47-2 | |
169.11 | |
LPUQAYUQRXPFSQ-UHFFFAOYNA-M | |
86748263 | |
[Na+].NC(CCC(O)=O)C([O-])=O |
C5H8NNaO4 | |
MFCD00150138 | |
natriumglutaminat, sodium glutamate, glutamate sodium, monosodioglutammato, sodium l-glutamate, glutammato monosodico, monosodium l-glutamate, sodium hydrogen glutamate, monosodium glutamate, glutamic acid, monosodium salt | |
sodium 2-amino-4-carboxybutanoate |
Safety and Handling
EINECSNumber : 205-538-1
Recommended Storage : Store at room temperature (15 to 30°C).