missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Glutamic acid dibenzyl ester p-toluenesulfonate, 98%

$138.65 - $1,378.65

Chemical Identifiers

CAS 2791-84-6
Molecular Formula C26H29NO7S
Molecular Weight (g/mol) 499.578
MDL Number MFCD00038980
InChI Key HVZUAIVKRYGQRM-LMOVPXPDSA-N
Synonym h-glu obzl-obzl.tosoh, l-glutamic acid dibenzyl ester tosylate, s-dibenzyl 2-aminopentanedioate 4-methylbenzenesulfonate, h-glu obzl-obzl p-tosylate, l-glutamic acid dibenzyl ester 4-toluenesulfonate, glu obzl-obzl.tosoh, dibenzyl l-glutamate p-toluenesulfonate, h-glu obzl-obzl 4-tosylate, l-glutamic acid dibenzyl ester p-toluenesulfonate, h-glu obzl-obzl inverted exclamation mark currencytosoh
PubChem CID 13233366
IUPAC Name dibenzyl (2S)-2-aminopentanedioate;4-methylbenzenesulfonic acid
SMILES CC1=CC=C(C=C1)S(=O)(=O)O.C1=CC=C(C=C1)COC(=O)CCC(C(=O)OCC2=CC=CC=C2)N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6269703 View Documents Alfa Aesar™
H6269703
1g Each for $138.65
 
AAH6269706 View Documents Alfa Aesar™
H6269706
5g Each for $618.39
 
AAH6269714 View Documents Alfa Aesar™
H6269714
25g Each for $1,378.65
 
Specifications

Chemical Identifiers

2791-84-6
499.578
HVZUAIVKRYGQRM-LMOVPXPDSA-N
13233366
CC1=CC=C(C=C1)S(=O)(=O)O.C1=CC=C(C=C1)COC(=O)CCC(C(=O)OCC2=CC=CC=C2)N
C26H29NO7S
MFCD00038980
h-glu obzl-obzl.tosoh, l-glutamic acid dibenzyl ester tosylate, s-dibenzyl 2-aminopentanedioate 4-methylbenzenesulfonate, h-glu obzl-obzl p-tosylate, l-glutamic acid dibenzyl ester 4-toluenesulfonate, glu obzl-obzl.tosoh, dibenzyl l-glutamate p-toluenesulfonate, h-glu obzl-obzl 4-tosylate, l-glutamic acid dibenzyl ester p-toluenesulfonate, h-glu obzl-obzl inverted exclamation mark currencytosoh
dibenzyl (2S)-2-aminopentanedioate;4-methylbenzenesulfonic acid

Specifications

2791-84-6
MFCD00038980
HVZUAIVKRYGQRM-LMOVPXPDSA-N
dibenzyl (2S)-2-aminopentanedioate;4-methylbenzenesulfonic acid
13233366
Powder
1g
C26H29NO7S
h-glu obzl-obzl.tosoh, l-glutamic acid dibenzyl ester tosylate, s-dibenzyl 2-aminopentanedioate 4-methylbenzenesulfonate, h-glu obzl-obzl p-tosylate, l-glutamic acid dibenzyl ester 4-toluenesulfonate, glu obzl-obzl.tosoh, dibenzyl l-glutamate p-toluenesulfonate, h-glu obzl-obzl 4-tosylate, l-glutamic acid dibenzyl ester p-toluenesulfonate, h-glu obzl-obzl inverted exclamation mark currencytosoh
CC1=CC=C(C=C1)S(=O)(=O)O.C1=CC=C(C=C1)COC(=O)CCC(C(=O)OCC2=CC=CC=C2)N
499.578
499.58
98%
L-Glutamic acid dibenzyl ester p-toluenesulfonate