missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Glutamic acid dibenzyl ester hydrochloride, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6359803
Description
L-Glutamic acid dibenzyl ester hydrochloride oxalate used as a intermediate in pharmaceutical and chemical synthesis. Also used in medicine research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsL-Glutamic acid dibenzyl ester hydrochloride oxalate used as a intermediate in pharmaceutical and chemical synthesis. Also used in medicine research.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
L-Glutamic acid dibenzyl ester hydrochloride | |
White | |
MFCD00035103 | |
GRGJVECUQLAEDM-LMOVPXPDSA-N | |
dibenzyl (2S)-2-aminopentanedioate;hydrochloride | |
199613 | |
98% | |
Powder |
4561-10-8 | |
C19H22ClNO4 | |
s-dibenzyl 2-aminopentanedioate hydrochloride, h-glu obzl-obzl.hcl, l-glutamic acid dibenzyl ester hydrochloride, l-glutamic acid, bis phenylmethyl ester, hydrochloride, s-dibenzyl 2-aminopentanedioate hcl, dibenzyl l-glutaminate hydrochloride, glutamic acid, dibenzyl ester, hydrochloride, l, 1,5-dibenzyl 2s-2-aminopentanedioate hydrochloride, h-glu obzl-obzl.hci, h-glu obzl-obzl hcl | |
C1=CC=C(C=C1)COC(=O)CCC(C(=O)OCC2=CC=CC=C2)N.Cl | |
363.84 | |
363.84 | |
1 g |
Chemical Identifiers
4561-10-8 | |
MFCD00035103 |
Safety and Handling
RTECSNumber : MA0885000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only