missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Glutamic acid di-tert-butyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6370814

 View more versions of this product

Catalog No. AAH6370814



Description

Description

L-Glutamic acid di-tert-butyl ester may be useful in the synthesis of substance P antagonistic peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Glutamic acid di-tert-butyl ester may be useful in the synthesis of substance P antagonistic peptides.

Solubility
Soluble in methanol (50gmg/ml).

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
Specifications

Specifications

L-Glutamic acid di-tert-butyl ester hydrochloride
C13H26ClNO4
3573738
Soluble in methanol (50gmg/ml).
CC(C)(C)OC(=O)CCC(C(=O)OC(C)(C)C)N.Cl
295.804
295.8
25 g
32677-01-3
MFCD00058003
l-glutamic acid di-tert-butyl ester hydrochloride, h-glu otbu-otbu.hcl, s-di-tert-butyl 2-aminopentanedioate hydrochloride, h-glu otbu-otbu hcl, l-glutamic acid, bis 1,1-dimethylethyl ester, hydrochloride, h-glu otbu-otbu inverted exclamation mark currencyhcl, glutamic acid di-t-butyl ester hydrochloride, s-di-tert-butyl 2-aminopentanedioate hcl, ditert-butyl 2s-2-aminopentanedioate hydrochloride, 1,5-di-tert-butyl 2s-2-aminopentanedioate hydrochloride
LFEYMWCCUAOUKZ-FVGYRXGTSA-N
ditert-butyl (2S)-2-aminopentanedioate;hydrochloride
208636
98%
Solid

Chemical Identifiers

32677-01-3
295.804
LFEYMWCCUAOUKZ-FVGYRXGTSA-N
208636
CC(C)(C)OC(=O)CCC(C(=O)OC(C)(C)C)N.Cl
C13H26ClNO4
MFCD00058003
l-glutamic acid di-tert-butyl ester hydrochloride, h-glu otbu-otbu.hcl, s-di-tert-butyl 2-aminopentanedioate hydrochloride, h-glu otbu-otbu hcl, l-glutamic acid, bis 1,1-dimethylethyl ester, hydrochloride, h-glu otbu-otbu inverted exclamation mark currencyhcl, glutamic acid di-t-butyl ester hydrochloride, s-di-tert-butyl 2-aminopentanedioate hcl, ditert-butyl 2s-2-aminopentanedioate hydrochloride, 1,5-di-tert-butyl 2s-2-aminopentanedioate hydrochloride
ditert-butyl (2S)-2-aminopentanedioate;hydrochloride
Safety and Handling

Safety and Handling

RTECSNumber : MA0880000

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only