Please login to your online account to display your discounted pricing

L-(+)-Glutamic Acid, BAKER ANALYZED™ Biochemical Reagent, J.T.Baker™

High quality biochemicals for research use

$217.41 - $366.41

Chemical Identifiers

Chemical Name or Material L-(+)-Glutamic Acid
CAS 56-86-0
Purity Grade BAKER ANALYZED™ Biochemical Reagent
Molecular Formula HOCOCH2CH2CH(NH2)COOH
Formula Weight 147.13
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003292

 
avantor performance
M75608
Poly Bottle 1kg Each for $366.41

02003293

 
avantor performance
M75607
Poly Bottle 500g Each for $217.41
Description & Specifications

Specifications

Formula Weight 147.13
Chemical Name or Material L-(+)-Glutamic Acid
Molecular Formula HOCOCH2CH2CH(NH2)COOH
CAS 56-86-0
Purity Grade BAKER ANALYZED™ Biochemical Reagent
Packaging Poly Bottle
Quantity 1kg
Specific Gravity 1L = 1.54kg