missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Glutamic acid 5-methyl ester, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 228311000

 View more versions of this product

Catalog No. AC228311000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

L-Glutamic acid 5-methyl ester
99%
(5% soln. in AcOH 80%) clear, colorless
≥98.5%
C6H11NO4
161.16
Authentic
+13° (24°C c=1,H2O)
100g
1499-55-4
Amorphous Powder
99%
CH3O2CCH2CH2CH(NH2)CO2H
MFCD00002632
+12° to +14° (24°C, 589nm) (c=1, H2O)
White

Chemical Identifiers

1499-55-4
C6H11NO4
161.16
L-Glutamic acid 5-methyl ester, 99%
99%
CH3O2CCH2CH2CH(NH2)CO2H
MFCD00002632
≥98.5%

Safety and Handling

SDS & Safety Documents

EINECSNumber : 216-110-9