Please login to your online account to display your discounted pricing

L-Glutamic acid 5-methyl ester, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material L-Glutamic acid 5-methyl ester
Name Note 99%
CAS 1499-55-4
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H11NO4
Linear Formula CH3O2CCH2CH2CH(NH2)CO2H
Formula Weight 161.16
MDL Number MFCD00002632
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC228311000

  SDS
thermo fisher scientific
228311000
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Formula Weight 161.16
Physical Form Amorphous Powder
Quantity 100g
Color White
Specific Rotation Condition +13° (24°C c=1,H2O)
EINECS Number 216-110-9
Appearance of Solution (5% soln. in AcOH 80%) clear, colorless
Infrared Spectrum Authentic
Specific Rotation +12° to +14° (24°C, 589nm) (c=1, H2O)
Chemical Name or Material L-Glutamic acid 5-methyl ester
MDL Number MFCD00002632
CAS 1499-55-4
Molecular Formula C6H11NO4
Name Note 99%
Assay Percent Range 99%
Linear Formula CH3O2CCH2CH2CH(NH2)CO2H