Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Glutamic acid 5-ethyl ester, 95%

Chemical Identifiers

Chemical Name or Material L-Glutamic acid 5-ethyl ester
CAS 1119-33-1
Formula Weight 175.18
MDL Number MFCD00037217
Synonym H-Glu(OEt)-OH; (S)-2-Amino-5-ethoxy-5-oxopentanoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6316806

 
alfa aesar™
H6316806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6316814

 
alfa aesar™
H6316814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 175.18
Chemical Name or Material L-Glutamic acid 5-ethyl ester
Physical Form Powder
Assay 95%
MDL Number MFCD00037217
CAS 1119-33-1
Synonym H-Glu(OEt)-OH; (S)-2-Amino-5-ethoxy-5-oxopentanoic acid
Color White to off-white
Melting Point 179°C (dec.)
Quantity 5g
Storage Note 1 2° to 8°C

Applications
L-Glutamic acid 5-ethyl ester is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water 100 mg/ml.

Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Safety and Handling

2° to 8°C