Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Glutamic acid 1-tert-butyl ester, 97%

Chemical Identifiers

Chemical Name or Material L-Glutamic acid 1-tert-butyl ester
CAS 45120-30-7
Formula Weight 203.24
MDL Number MFCD00038562
Beilstein 2414793
Synonym H-Glu-OtBu; (S)-4-Amino-5-(tert-butoxy)-5-oxopentanoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6534206

  SDS
alfa aesar™
H6534206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6534203

  SDS
alfa aesar™
H6534203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH65342MD

  SDS
alfa aesar™
H65342MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 203.24
Melting Point 139° to 145°C
MDL Number MFCD00038562
CAS 45120-30-7
Physical Form Powder
Assay 97%
Beilstein 2414793
Synonym H-Glu-OtBu; (S)-4-Amino-5-(tert-butoxy)-5-oxopentanoic acid
Chemical Name or Material L-Glutamic acid 1-tert-butyl ester
Color White
Quantity 5g
Storage Note 1 2° to 8°C
Safety and Handling

2° to 8°C