Please login to your online account to display your discounted pricing

L(-)-Galactose, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material L(-)-Galactose
CAS 15572-79-9
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H12O6
Formula Weight 180.16
MDL Number MFCD00063833
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC257971000

  SDS
thermo fisher scientific
AC257971000
Glass bottle 100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Fine Crystalline Powder
Assay ≥98.5%
Formula Weight 180.16
Quantity 100mg
Color White
Appearance of Solution (10% aq. soln.) clear, colorless
Assay Percent Range 99%
CAS 15572-79-9
Packaging Glass bottle
Specific Rotation -79° to -81° (20°C, 589nm) (c=5, water)
Chemical Name or Material L(-)-Galactose
Molecular Formula C6H12O6
MDL Number MFCD00063833
Infrared Spectrum Authentic