Please login to your online account to display your discounted pricing

L(-)-Galactose, 99%, ACROS Organics™


Quantity 100mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC257971000

 View more versions of this product

Catalog No. AC257971000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Fine Crystalline Powder
Assay ≥98.5%
Formula Weight 180.16
Quantity 100mg
Color White
Appearance of Solution (10% aq. soln.) clear, colorless
Assay Percent Range 99%
CAS 15572-79-9
Packaging Glass bottle
Specific Rotation -79° to -81° (20°C, 589nm) (c=5, water)
Chemical Name or Material L(-)-Galactose
Molecular Formula C6H12O6
MDL Number MFCD00063833
Infrared Spectrum Authentic

Specifications

Chemical Name or Material L(-)-Galactose
CAS 15572-79-9
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H12O6
Formula Weight 180.16
MDL Number MFCD00063833