missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-(+)-Fructose, TCI America™

Manufacturer:  TCI America F03171G

 View more versions of this product

Catalog No. F03171G



Specifications

Specifications

L-(+)-Fructose
1g
C6H12O6
BJHIKXHVCXFQLS-FUTKDDECSA-N
(3R,4S,5S)-1,3,4,5,6-pentahydroxyhexan-2-one
5460024
180.16
≥95.0% (HPLC)
White
7776-48-9
MFCD05662378
C(C(C(C(C(=O)CO)O)O)O)O
180.156
CHEBI:37724
Crystal-Powder at 20°C

Chemical Identifiers

7776-48-9
180.156
BJHIKXHVCXFQLS-FUTKDDECSA-N
CHEBI:37724
C(C(C(C(C(=O)CO)O)O)O)O
C6H12O6
MFCD05662378
5460024
(3R,4S,5S)-1,3,4,5,6-pentahydroxyhexan-2-one
Safety and Handling

Safety and Handling

TSCA : No

Documents