missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Cystine disodium salt, 98+%

$137.24 - $530.49

Chemical Identifiers

CAS 64704-23-0
Molecular Formula C6H10N2Na2O4S2
Molecular Weight (g/mol) 284.256
MDL Number MFCD00037206
InChI Key PLVPMKWGXOOSKL-VBCXBIGUSA-L
Synonym disodium 2-amino-3-2r-2-amino-2-carboxylatoethyl disulfanyl propanoate
PubChem CID 71607258
IUPAC Name disodium;2-amino-3-[[(2R)-2-amino-2-carboxylatoethyl]disulfanyl]propanoate
SMILES C(C(C(=O)[O-])N)SSCC(C(=O)[O-])N.[Na+].[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6231030 View Documents Alfa Aesar™
J6231030
250g Each for $137.24
 
AAJ62310A1 View Documents Alfa Aesar™
J62310A1
1kg Each for $530.49
 
Specifications

Chemical Identifiers

64704-23-0
284.256
PLVPMKWGXOOSKL-VBCXBIGUSA-L
71607258
C(C(C(=O)[O-])N)SSCC(C(=O)[O-])N.[Na+].[Na+]
C6H10N2Na2O4S2
MFCD00037206
disodium 2-amino-3-2r-2-amino-2-carboxylatoethyl disulfanyl propanoate
disodium;2-amino-3-[[(2R)-2-amino-2-carboxylatoethyl]disulfanyl]propanoate

Specifications

+170° (c=2 in 1N HCl)
250g
C6H10N2Na2O4S2
disodium 2-amino-3-2r-2-amino-2-carboxylatoethyl disulfanyl propanoate
C(C(C(=O)[O-])N)SSCC(C(=O)[O-])N.[Na+].[Na+]
284.256
284.3
≥98%
Brown
64704-23-0
MFCD00037206
PLVPMKWGXOOSKL-VBCXBIGUSA-L
disodium;2-amino-3-[[(2R)-2-amino-2-carboxylatoethyl]disulfanyl]propanoate
71607258
Powder
L-Cystine disodium salt
Safety and Handling

Safety and Handling

TSCA : TSCA