missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Cystine Dimethyl Ester Dihydrochloride, TCI America™

Manufacturer:  TCI America C330325G

 View more versions of this product

Catalog No. C330325G



Specifications

Specifications

L-Cystine Dimethyl Ester Dihydrochloride
179°C
32854-09-4
MFCD00012490
QKWGUPFPCRKKMQ-BNTLRKBRSA-N
methyl (2S)-2-amino-3-[[(2S)-2-amino-3-methoxy-3-oxopropyl]disulfanyl]propanoate;dihydrochloride
71463656
Crystal-Powder at 20°C
White
25g
C8H18Cl2N2O4S2
h-cys-ome 2.2hcl, dimethyl l-cystinate dihydrochloride, h-d-cys-ome 2 2hcl, methyl 2-amino-3-r-2-amino-3-methoxy-3-oxopropyl disulfanyl propanoate dihydrochloride, d-cystine bis methyl ester dihydrochloride, d-cystine dimethyl ester dihydrochloride, h-d-cys-ome 2 hcl, c8h16n2o4s2.2hcl, dimethyl l-cystinate 2hcl, h-d-cys-ome 2 . 2 hcl
COC(=O)C(CSSCC(C(=O)OC)N)N.Cl.Cl
341.262
341.26
≥96.0% (T)

Chemical Identifiers

32854-09-4
341.262
QKWGUPFPCRKKMQ-BNTLRKBRSA-N
71463656
COC(=O)C(CSSCC(C(=O)OC)N)N.Cl.Cl
C8H18Cl2N2O4S2
MFCD00012490
h-cys-ome 2.2hcl, dimethyl l-cystinate dihydrochloride, h-d-cys-ome 2 2hcl, methyl 2-amino-3-r-2-amino-3-methoxy-3-oxopropyl disulfanyl propanoate dihydrochloride, d-cystine bis methyl ester dihydrochloride, d-cystine dimethyl ester dihydrochloride, h-d-cys-ome 2 hcl, c8h16n2o4s2.2hcl, dimethyl l-cystinate 2hcl, h-d-cys-ome 2 . 2 hcl
methyl (2S)-2-amino-3-[[(2S)-2-amino-3-methoxy-3-oxopropyl]disulfanyl]propanoate;dihydrochloride
Safety and Handling

Safety and Handling

TSCA : No

Documents