Please login to your online account to display your discounted pricing

L-Cysteine Hydrochloride, Monohydrate, Multi-Compendial, U.S.P., J.T.Baker™


Quantity 1kg
Packaging Poly Bottle

Manufacturer: avantor performance  207106

 View more versions of this product

Catalog No. 02003184

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 175.64
Purity Grade Multi-Compendial/USP
Chemical Name or Material L-Cysteine Hydrochloride
CAS 7048-04-6
Molecular Formula C3H7NO2S·HCl·H2O
Name Note Monohydrate
Specific Gravity 1L = 1.50kg
Packaging Poly Bottle
Quantity 1kg

Specifications

Chemical Name or Material L-Cysteine Hydrochloride
CAS 7048-04-6
Name Note Monohydrate
Purity Grade Multi-Compendial/USP
Molecular Formula C3H7NO2S·HCl·H2O
Formula Weight 175.64