Please login to your online account to display your discounted pricing

L-(+)-Cysteine Hydrochloride, Monohydrate, BAKER ANALYZED™ Biochemical Reagent, J.T.Baker™


High quality biochemicals for research use

Quantity 100g
Packaging Wide Mouth Amber Glass Bottle

Manufacturer: avantor performance  G12105

 View more versions of this product

Catalog No. 02003150

  • / Case of 4 Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 175.64
Specific Gravity 1L = 1.50kg
Chemical Name or Material L-(+)-Cysteine Hydrochloride
Name Note Monohydrate
Molecular Formula HSCH2CH(NH2)COOH·HCl·H2O
CAS 7048-04-6
Purity Grade BAKER ANALYZED™ Biochemical Reagent
Packaging Wide Mouth Amber Glass Bottle
Quantity 100g

Specifications

Chemical Name or Material L-(+)-Cysteine Hydrochloride
CAS 7048-04-6
Name Note Monohydrate
Purity Grade BAKER ANALYZED™ Biochemical Reagent
Molecular Formula HSCH2CH(NH2)COOH·HCl·H2O
Formula Weight 175.64