Please login to your online account to display your discounted pricing

L-(+)-Cysteine Hydrochloride, Monohydrate, BAKER ANALYZED™ Biochemical Reagent, J.T.Baker™

High quality biochemicals for research use

$112.19 - $112.19

Chemical Identifiers

Chemical Name or Material L-(+)-Cysteine Hydrochloride
CAS 7048-04-6
Name Note Monohydrate
Purity Grade BAKER ANALYZED™ Biochemical Reagent
Molecular Formula HSCH2CH(NH2)COOH·HCl·H2O
Formula Weight 175.64
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003151

 
avantor performance
G12103
Wide Mouth Amber Glass Bottle 25g Each for $112.19
Description & Specifications

Specifications

Formula Weight 175.64
CAS 7048-04-6
Chemical Name or Material L-(+)-Cysteine Hydrochloride
Molecular Formula HSCH2CH(NH2)COOH·HCl·H2O
Purity Grade BAKER ANALYZED™ Biochemical Reagent
Name Note Monohydrate
Packaging Wide Mouth Amber Glass Bottle
Quantity 25g
Specific Gravity 1L = 1.50kg