Please login to your online account to display your discounted pricing

L-(+)-Cysteine Hydrochloride, Monohydrate, BAKER ANALYZED™ Biochemical Reagent, J.T.Baker™


High quality biochemicals for research use

Quantity 25g
Packaging Wide Mouth Amber Glass Bottle

Manufacturer: avantor performance  G12103

 View more versions of this product

Catalog No. 02003151

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 175.64
CAS 7048-04-6
Chemical Name or Material L-(+)-Cysteine Hydrochloride
Molecular Formula HSCH2CH(NH2)COOH·HCl·H2O
Purity Grade BAKER ANALYZED™ Biochemical Reagent
Name Note Monohydrate
Packaging Wide Mouth Amber Glass Bottle
Quantity 25g
Specific Gravity 1L = 1.50kg

Specifications

Chemical Name or Material L-(+)-Cysteine Hydrochloride
CAS 7048-04-6
Name Note Monohydrate
Purity Grade BAKER ANALYZED™ Biochemical Reagent
Molecular Formula HSCH2CH(NH2)COOH·HCl·H2O
Formula Weight 175.64