Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Cysteic acid monohydrate, 99%

$232.04 - $232.04

Chemical Identifiers

Chemical Name or Material L-Cysteic acid monohydrate
CAS 23537-25-9
Molecular Formula C3H7NO5S·H2O
Formula Weight 187.19 (169.17 Anhydrous)
MDL Number MFCD00149544
Beilstein 1725495
Merck Index 14,2780
Synonym 2-Amino-3-sulfopropionic acid monohydrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0913514

  SDS
alfa aesar™
L0913514
25g Each for $232.04
Description & Specifications

Specifications

Formula Weight 187.19 (169.17 Anhydrous)
Chemical Name or Material L-Cysteic acid monohydrate
CAS 23537-25-9
MDL Number MFCD00149544
Molecular Formula C3H7NO5S·H2O
Assay 99%
Synonym 2-Amino-3-sulfopropionic acid monohydrate
Quantity 25g
Optical Rotation +8° (c=5 in Water)
Melting Point 268°C
Beilstein 1725495
Merck Index 14,2780

Applications
L-Cysteic acid monohydrate is used as a competitive inhibitor of the bacterial aspartate:alanine antiporter (AspT) exchange of aspartate and in other aspartate biological systems. It is used in monomeric surfactant development. It is used in studies of excitatory amino acids in the brain, such as those that bind to cysteine sulfinic acid receptors. L-Cysteic acid is a useful agonist at several rat metabotropic glutamate receptors (mGluRs).

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents. Store away from oxidizing agents.