missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Citrulline, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1331618
Description
L-Citrulline, is used as an essential intermediate in the biosynthesis of nitric oxide from L-arginine . It is also used as a nutritional drink and biochemical reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsL-Citrulline, is used as an essential intermediate in the biosynthesis of nitric oxide from L-arginine . It is also used as a nutritional drink and biochemical reagent.
Solubility
It is soluble in water.
Notes
Stable under ordinary conditions. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents.
Specifications
L-Citrulline | |
212°C to 214°C (decomposition) | |
MFCD00064397 | |
6055157 | |
l-citrulline, citrulline, h-cit-oh, delta-ureidonorvaline, sitrulline, n5-carbamoyl-l-ornithine, l-cytrulline, n5-aminocarbonyl ornithine, n delta-carbamylornithine, citrulline, l | |
RHGKLRLOHDJJDR-UHFFFAOYNA-N | |
NC(CCCNC(N)=O)C(O)=O | |
175.19 | |
CHEBI:16349 | |
98% |
372-75-8 | |
C6H13N3O3 | |
50 g | |
14,2331 | |
It is soluble in water. | |
+22° (c=2 in 1N HCl) | |
(2S)-2-amino-5-(carbamoylamino)pentanoic acid | |
9750 | |
175.19 |
Chemical Identifiers
372-75-8 | |
175.19 | |
RHGKLRLOHDJJDR-UHFFFAOYNA-N | |
9750 | |
(2S)-2-amino-5-(carbamoylamino)pentanoic acid |
C6H13N3O3 | |
MFCD00064397 | |
l-citrulline, citrulline, h-cit-oh, delta-ureidonorvaline, sitrulline, n5-carbamoyl-l-ornithine, l-cytrulline, n5-aminocarbonyl ornithine, n delta-carbamylornithine, citrulline, l | |
CHEBI:16349 | |
NC(CCCNC(N)=O)C(O)=O |
Safety and Handling
EINECSNumber : 206-759-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only