missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L(-)-Camphorsulfonic acid, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC263645000

 View more versions of this product

Catalog No. AC263645000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

L(-)-Camphorsulfonic acid
Glass bottle
White to Beige
500g
98%
MFCD00064158
16,58
Solubility in water: soluble. Other solubilities: slightly soluble in glacial acetic acid and ethyl,acetate,practically insoluble in ether,soluble in chloroform and alcohol
CC1(C2CCC1(C(=O)C2)CS(=O)(=O)O)C
232.3
CHEBI:55401
Crystalline Powder
Authentic
− 22.00
198.0°C
35963-20-3
C10H16O4S
11,316
--camphor-10-sulfonic acid, 1r,4s-7,7-dimethyl-2-oxobicyclo 2.2.1 hept-1-yl methanesulfonic acid, 1r,4s-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonic acid, 1r---camphor-10-sulfonic acid, camphorsulfonic acid,-, l---camphor-10-sulfonic acid, l--camphorsulfonic acid, s-camphorsulfonic acid, unii-y6075i4fxe
MIOPJNTWMNEORI-XVKPBYJWSA-N
[(1S,4R)-7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl]methanesulfonic acid
5771688
232.3
98%

Chemical Identifiers

35963-20-3
232.3
MIOPJNTWMNEORI-XVKPBYJWSA-N
5771688
[(1S,4R)-7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl]methanesulfonic acid
C10H16O4S
MFCD00064158
--camphor-10-sulfonic acid, 1r,4s-7,7-dimethyl-2-oxobicyclo 2.2.1 hept-1-yl methanesulfonic acid, 1r,4s-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonic acid, 1r---camphor-10-sulfonic acid, camphorsulfonic acid,-, l---camphor-10-sulfonic acid, l--camphorsulfonic acid, s-camphorsulfonic acid, unii-y6075i4fxe
CHEBI:55401
CC1(C2CCC1(C(=O)C2)CS(=O)(=O)O)C
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 252-817-9

SDS Documents