Please login to your online account to display your discounted pricing

L(-)-Camphor, ACROS Organics™

$142.63 - $142.63

Chemical Identifiers

Chemical Name or Material L(-)-Camphor
CAS 464-48-2
Purity Grade Technical Grade
Assay ≥85%
Molecular Formula C10H16O
Formula Weight 152.24
MDL Number MFCD00064148
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC108190100

  SDS
thermo fisher scientific
108190100
Glass bottle 10g Each for $142.63
Description & Specifications

Specifications

Assay ≥85%
Formula Weight 152.24
Purity Grade Technical Grade
Physical Form Adhering Crystals or Crystalline Powder
Quantity 10g
Color White to yellow
Beilstein 07, 134
Merck Index 15, 1734
Specific Rotation -28° to -32° (20°C, 589nm) (c=10, alcohol)
Chemical Name or Material L(-)-Camphor
Molecular Formula C10H16O
CAS 464-48-2
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Flammable solid.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00064148
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Warning
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Flammable solid.