missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Buthionine-(S,R)-sulfoximine, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 235520050

 View more versions of this product

Catalog No. AC235520050



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

L-Buthionine-(S, R)-sulfoximine
234.0°C to 235.0°C
Authentic
Glass bottle
HOOCCH(NH2)CH2CH2S(O)(=NH)(CH2)3CH3
5 g
+ 33.00
Solubility in water: soluble
CCCCS(=N)(=O)CCC(C(=O)O)N
222.31
222.31
Fine Powder
83730-53-4
White
99%
C8H18N2O3S
MFCD00067000
+33° (20°C c=1,1N HCl)
l-buthionine-sulfoximine, l-buthionine sulfoximine, 2s-2-amino-4-butylsulfonimidoyl butanoic acid, l-buthionine-s,r-sulfoximine, l-buthionine s,r-sulfoximine, l-bso, butanoic acid, 2-amino-4-s-butylsulfonimidoyl-, 2s, l-buthionine sr-sulfoximine, buthionine-s,r-sulfoximine, l, l-buthionine-s,r-sulphoximine
KJQFBVYMGADDTQ-CVSPRKDYSA-N
(2S)-2-amino-4-(butylsulfonimidoyl)butanoic acid
119565
99%

Chemical Identifiers

83730-53-4
222.31
KJQFBVYMGADDTQ-CVSPRKDYSA-N
119565
CCCCS(=N)(=O)CCC(C(=O)O)N
C8H18N2O3S
MFCD00067000
l-buthionine-sulfoximine, l-buthionine sulfoximine, 2s-2-amino-4-butylsulfonimidoyl butanoic acid, l-buthionine-s,r-sulfoximine, l-buthionine s,r-sulfoximine, l-bso, butanoic acid, 2-amino-4-s-butylsulfonimidoyl-, 2s, l-buthionine sr-sulfoximine, buthionine-s,r-sulfoximine, l, l-buthionine-s,r-sulphoximine
(2S)-2-amino-4-(butylsulfonimidoyl)butanoic acid
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only