missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Bornyl acetate, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC106550025

 View more versions of this product

Catalog No. AC106550025

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Liquid
196.29
Clear colorless
0.986 to 0.99 (15°/15°C)
1.4620 to 1.4650 (20°C, 589nm)
95%
C12H20O2
MFCD00867808
95%
≥94%
2.5kg
0.9875g/mL
− 38.00 (20.00°C c=neat)
Authentic
(20°C, 589nm) (neat) < -38°
L-Bornyl acetate
5655-61-8
total esters (GC):> 98% (as C12H20O2)

Chemical Identifiers

5655-61-8
C12H20O2
MFCD00867808
≥94%
95%
196.29
L-Bornyl acetate, 95%

Safety and Handling

SDS & Safety Documents

EINECSNumber : 227-101-4

TSCA : TSCA