Please login to your online account to display your discounted pricing

Alfa Aesar™ L-beta-Homoproline hydrochloride, 98%

Chemical Identifiers

Chemical Name or Material L-beta-Homoproline hydrochloride
CAS 53912-85-9
Formula Weight 165.62
MDL Number MFCD07363484
Beilstein 471672
Synonym L-β-HomoPro.HCl; (S)-2-(2-Pyrrolidinyl)acetic acid hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6511003

  SDS
alfa aesar™
H6511003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH65110MD

  SDS
alfa aesar™
H65110MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 165.62
Chemical Name or Material L-beta-Homoproline hydrochloride
CAS 53912-85-9
MDL Number MFCD07363484
Synonym L-β-HomoPro.HCl; (S)-2-(2-Pyrrolidinyl)acetic acid hydrochloride
Assay 98%
Color White
Physical Form Powder
Quantity 1g
Sensitivity Hygroscopic
Beilstein 471672