missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Aspartic acid monosodium salt monohydrate, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC451620010

 View more versions of this product

Catalog No. AC451620010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

L-Aspartic acid monosodium salt monohydrate
97%
monosodium l-aspartate dihydrate
VJDUMAWJHIHDHJ-SQGDDOFFSA-N
(2S)-2-aminobutanedioic acid;sodium;dihydrate
129893661
97%
3792-50-5
C4H11NNaO6
Plastic bottle
C(C(C(=O)O)N)C(=O)O.O.O.[Na]
192.123
173.1
1kg

Chemical Identifiers

3792-50-5
192.123
monosodium l-aspartate dihydrate
(2S)-2-aminobutanedioic acid;sodium;dihydrate
C4H11NNaO6
VJDUMAWJHIHDHJ-SQGDDOFFSA-N
129893661
C(C(C(=O)O)N)C(=O)O.O.O.[Na]
SDS Documents