missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Aspartic acid monosodium salt monohydrate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 451620250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
L-Aspartic acid monosodium salt monohydrate | |
97% | |
Plastic Bottle | |
VJDUMAWJHIHDHJ-SQGDDOFFSA-N | |
(2S)-2-aminobutanedioic acid;sodium;dihydrate | |
129893661 | |
97% |
3792-50-5 | |
C4H11NNaO6 | |
monosodium l-aspartate dihydrate | |
C(C(C(=O)O)N)C(=O)O.O.O.[Na] | |
192.123 | |
173.1 | |
25 g |
Chemical Identifiers
3792-50-5 | |
192.123 | |
monosodium l-aspartate dihydrate | |
(2S)-2-aminobutanedioic acid;sodium;dihydrate |
C4H11NNaO6 | |
VJDUMAWJHIHDHJ-SQGDDOFFSA-N | |
129893661 | |
C(C(C(=O)O)N)C(=O)O.O.O.[Na] |
RUO – Research Use Only