Please login to your online account to display your discounted pricing

L-aspartic acid dimethyl ester hydrochloride, 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC376660050

 View more versions of this product

Catalog No. AC376660050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 197.62
Physical Form Crystalline Powder
Quantity 5g
Color White to off-white
Specific Rotation Condition +15° (20°C c=1,Chloroform)
Packaging Glass bottle
CAS 32213-95-9
Assay Percent Range 98%
MDL Number MFCD00038878
Specific Rotation +13° to +17° (20°C, 589nm) (c=1, CH3OH)
Infrared Spectrum Authentic
Linear Formula CH3OC(O)CH2CH(NH2)C(O)OCH3·HCl
Chemical Name or Material L-Aspartic acid dimethyl ester hydrochloride
Molecular Formula C6H11NO4·HCl

Specifications

Chemical Name or Material L-Aspartic acid dimethyl ester hydrochloride
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H11NO4·HCl
Linear Formula CH3OC(O)CH2CH(NH2)C(O)OCH3·HCl
Formula Weight 197.62
MDL Number MFCD00038878