missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Aspartic acid diethyl ester hydrochloride, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6397414
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
L-Aspartic acid diethyl ester hydrochloride | |
C8H16ClNO4 | |
l-aspartic acid diethyl ester hydrochloride, h-asp oet-oet.hcl, h-asp oet-oet hcl, s-diethyl 2-aminosuccinate hydrochloride, aspartic acid, diethyl ester, hydrochloride, h-asp oet-oet hydrochloride, diethyl l-aspartate hydrochloride, 1,4-diethyl 2s-2-aminobutanedioate hydrochloride, h-asp oet-oet?cl, diethylaspartate hydrochloride | |
CCOC(=O)CC(C(=O)OCC)N.Cl | |
225.669 | |
225.67 | |
25 g |
16115-68-7 | |
MFCD00038910 | |
AJOXZAAREAYBQR-RGMNGODLSA-N | |
diethyl (2S)-2-aminobutanedioate;hydrochloride | |
12416466 | |
98% | |
Solid |
Chemical Identifiers
16115-68-7 | |
225.669 | |
AJOXZAAREAYBQR-RGMNGODLSA-N | |
12416466 | |
CCOC(=O)CC(C(=O)OCC)N.Cl |
C8H16ClNO4 | |
MFCD00038910 | |
l-aspartic acid diethyl ester hydrochloride, h-asp oet-oet.hcl, h-asp oet-oet hcl, s-diethyl 2-aminosuccinate hydrochloride, aspartic acid, diethyl ester, hydrochloride, h-asp oet-oet hydrochloride, diethyl l-aspartate hydrochloride, 1,4-diethyl 2s-2-aminobutanedioate hydrochloride, h-asp oet-oet?cl, diethylaspartate hydrochloride | |
diethyl (2S)-2-aminobutanedioate;hydrochloride |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only