missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Aspartic acid diethyl ester hydrochloride, 98%

Manufacturer:  Alfa Aesar™ H6397422

 View more versions of this product

Catalog No. AAH6397422



Specifications

Specifications

L-Aspartic acid diethyl ester hydrochloride
C8H16ClNO4
l-aspartic acid diethyl ester hydrochloride, h-asp oet-oet.hcl, h-asp oet-oet hcl, s-diethyl 2-aminosuccinate hydrochloride, aspartic acid, diethyl ester, hydrochloride, h-asp oet-oet hydrochloride, diethyl l-aspartate hydrochloride, 1,4-diethyl 2s-2-aminobutanedioate hydrochloride, h-asp oet-oet?cl, diethylaspartate hydrochloride
CCOC(=O)CC(C(=O)OCC)N.Cl
225.669
225.67
98%
16115-68-7
MFCD00038910
AJOXZAAREAYBQR-RGMNGODLSA-N
diethyl (2S)-2-aminobutanedioate;hydrochloride
12416466
Solid
100g

Chemical Identifiers

16115-68-7
225.669
AJOXZAAREAYBQR-RGMNGODLSA-N
12416466
CCOC(=O)CC(C(=O)OCC)N.Cl
C8H16ClNO4
MFCD00038910
l-aspartic acid diethyl ester hydrochloride, h-asp oet-oet.hcl, h-asp oet-oet hcl, s-diethyl 2-aminosuccinate hydrochloride, aspartic acid, diethyl ester, hydrochloride, h-asp oet-oet hydrochloride, diethyl l-aspartate hydrochloride, 1,4-diethyl 2s-2-aminobutanedioate hydrochloride, h-asp oet-oet?cl, diethylaspartate hydrochloride
diethyl (2S)-2-aminobutanedioate;hydrochloride
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C

SDS Documents