missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Aspartic acid dibenzyl ester p-toluenesulfonate, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6562314
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
157°C to 160°C | |
2886-33-1 | |
C25H27NO7S | |
3647510 | |
HLMUYZYLPUHSNV-UHFFFAOYNA-N | |
1,4-dibenzyl 2-aminobutanedioate; 4-methylbenzene-1-sulfonic acid | |
2808531 | |
95% | |
Powder |
L-Aspartic acid dibenzyl ester p-toluenesulfonate | |
Brown | |
MFCD00065188 | |
h-asp obzl-obzl.tosoh, h-asp obzl-obzl tosoh, l-aspartic acid dibenzyl ester tosylate, s-dibenzyl 2-aminosuccinate 4-methylbenzenesulfonate, l-aspartic acid dibenzyl ester p-toluenesulfonate salt, l-aspartic acid dibenzyl ester 4-toluenesulfonate, h-asp obzl-obzl inverted exclamation mark currencytosoh, h-asp obzl-obzl.tos, l-aspartic acid dibenzyl ester p-toluenesulfonate, 1,4-dibenzyl 2s-2-aminobutanedioate; para-toluene sulfonate | |
CC1=CC=C(C=C1)S(O)(=O)=O.NC(CC(=O)OCC1=CC=CC=C1)C(=O)OCC1=CC=CC=C1 | |
485.55 | |
485.55 | |
25 g |
Chemical Identifiers
2886-33-1 | |
485.55 | |
HLMUYZYLPUHSNV-UHFFFAOYNA-N | |
2808531 | |
CC1=CC=C(C=C1)S(O)(=O)=O.NC(CC(=O)OCC1=CC=CC=C1)C(=O)OCC1=CC=CC=C1 |
C25H27NO7S | |
MFCD00065188 | |
h-asp obzl-obzl.tosoh, h-asp obzl-obzl tosoh, l-aspartic acid dibenzyl ester tosylate, s-dibenzyl 2-aminosuccinate 4-methylbenzenesulfonate, l-aspartic acid dibenzyl ester p-toluenesulfonate salt, l-aspartic acid dibenzyl ester 4-toluenesulfonate, h-asp obzl-obzl inverted exclamation mark currencytosoh, h-asp obzl-obzl.tos, l-aspartic acid dibenzyl ester p-toluenesulfonate, 1,4-dibenzyl 2s-2-aminobutanedioate; para-toluene sulfonate | |
1,4-dibenzyl 2-aminobutanedioate; 4-methylbenzene-1-sulfonic acid |
Safety and Handling
EINECSNumber : 220-746-2
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only