Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Aspartic acid dibenzyl ester hydrochloride, 97%

Chemical Identifiers

Chemical Name or Material L-Aspartic acid dibenzyl ester hydrochloride
CAS 6327-59-9
Formula Weight 349.81
MDL Number MFCD01075101
Beilstein 3921431
Synonym H-Asp(OBzl)-OBzl.HCl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6535306

  SDS
alfa aesar™
H6535306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6535314

  SDS
alfa aesar™
H6535314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6535322

  SDS
alfa aesar™
H6535322
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 349.81
CAS 6327-59-9
MDL Number MFCD01075101
Assay 97%
Chemical Name or Material L-Aspartic acid dibenzyl ester hydrochloride
Synonym H-Asp(OBzl)-OBzl.HCl
Physical Form Solid
Beilstein 3921431
Quantity 5g
Storage Note 1 2° to 8°C
Safety and Handling

2° to 8°C