missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Aspartic acid di-tert-butyl ester hydrochloride, 98%

Manufacturer:  Alfa Aesar™ H6371614

 View more versions of this product

Catalog No. AAH6371614



Description

Description

Applications
L-Aspartic acid di-tert-butyl ester hydrochloride may be used in neurological comparison studies with structurally similar compounds such as L-Glutamic acid di-tert-butyl ester.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
Specifications

Specifications

1791-13-5
MFCD00034851
1,4-di-tert-butyl 2s-2-aminobutanedioate hydrochloride, di-tert-butyl l-aspartate hydrochloride, h-asp obut-obut·hcl, h-asp otbu-otbu hcl, h-asp otbu-otbu.hcl, h-asp otbu-otbucl, l-aspartic acid di-t-butyl ester hydrochloride, l-aspartic acid di-tert-butyl ester hydrochloride, l-aspartic acid, bis 1,1-dimethylethyl ester, hydrochloride, s-di-tert-butyl 2-aminosuccinate hydrochloride
CC(C)(C)OC(=O)CC(C(=O)OC(C)(C)C)N.Cl
281.777
281.78
98%
C12H24ClNO4
5652807
GVLZIMQSYQDAHB-QRPNPIFTSA-N
ditert-butyl (2S)-2-aminobutanedioate;hydrochloride
13768146
Solid
25g

Chemical Identifiers

1791-13-5
281.777
GVLZIMQSYQDAHB-QRPNPIFTSA-N
13768146
CC(C)(C)OC(=O)CC(C(=O)OC(C)(C)C)N.Cl
C12H24ClNO4
MFCD00034851
1,4-di-tert-butyl 2s-2-aminobutanedioate hydrochloride, di-tert-butyl l-aspartate hydrochloride, h-asp obut-obut·hcl, h-asp otbu-otbu hcl, h-asp otbu-otbu.hcl, h-asp otbu-otbucl, l-aspartic acid di-t-butyl ester hydrochloride, l-aspartic acid di-tert-butyl ester hydrochloride, l-aspartic acid, bis 1,1-dimethylethyl ester, hydrochloride, s-di-tert-butyl 2-aminosuccinate hydrochloride
ditert-butyl (2S)-2-aminobutanedioate;hydrochloride
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

Documents