missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Aspartic acid, 99%, low metals content

Manufacturer:  Alfa Aesar™ 4331722

 View more versions of this product

Catalog No. AA4331722



Specifications

Specifications

L-Aspartic acid
56-84-8
MFCD00002616
14,840
CKLJMWTZIZZHCS-REOHCLBHSA-N
(2S)-2-aminobutanedioic acid
5960
133.1
99%
>300°C (decomposition)
Odorless
Low metals content
C4H7NO4
1723530
asparagic acid
C(C(C(=O)O)N)C(=O)O
133.103
CHEBI:17053
Powder
1.66g/mL
100g

Chemical Identifiers

56-84-8
133.103
CKLJMWTZIZZHCS-REOHCLBHSA-N
5960
(2S)-2-aminobutanedioic acid
C4H7NO4
MFCD00002616
asparagic acid
CHEBI:17053
C(C(C(=O)O)N)C(=O)O
Safety and Handling

Safety and Handling

RTECSNumber : CI9098500

TSCA : TSCA

Documents