missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Aspartic acid 4-tert-butyl 1-methyl ester hydrochloride, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6325706
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
L-Aspartic acid 4-tert-butyl 1-methyl ester hydrochloride | |
C9H18ClNO4 | |
6005591 | |
SFYKWYAIJZEDNG-RGMNGODLSA-N | |
4-O-tert-butyl 1-O-methyl (2S)-2-aminobutanedioate;hydrochloride | |
13970777 | |
98% | |
Solid |
2673-19-0 | |
MFCD00153427 | |
h-asp otbu-ome.hcl, h-asp otbu-ome hcl, l-aspartic acid 4-tert-butyl-1-methyl ester hydrochloride, h-asp otbu-ome hydrochloride, h-asp obut-ome hcl, s-4-tert-butyl 1-methyl 2-aminosuccinate hydrochloride, 4-tert-butyl 1-methyl 2s-2-aminobutanedioate hydrochloride, h-asp otbu-ome, h-asp otbu-omecl, h-asp o-tbu-ome | |
CC(C)(C)OC(=O)CC(C(=O)OC)N.Cl | |
239.696 | |
239.7 | |
5 g |
Chemical Identifiers
2673-19-0 | |
239.696 | |
SFYKWYAIJZEDNG-RGMNGODLSA-N | |
13970777 | |
CC(C)(C)OC(=O)CC(C(=O)OC)N.Cl |
C9H18ClNO4 | |
MFCD00153427 | |
h-asp otbu-ome.hcl, h-asp otbu-ome hcl, l-aspartic acid 4-tert-butyl-1-methyl ester hydrochloride, h-asp otbu-ome hydrochloride, h-asp obut-ome hcl, s-4-tert-butyl 1-methyl 2-aminosuccinate hydrochloride, 4-tert-butyl 1-methyl 2s-2-aminobutanedioate hydrochloride, h-asp otbu-ome, h-asp otbu-omecl, h-asp o-tbu-ome | |
4-O-tert-butyl 1-O-methyl (2S)-2-aminobutanedioate;hydrochloride |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only