missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Aspartic acid 4-methyl ester hydrochloride, 97%

Manufacturer:  Alfa Aesar™ H6217206

 View more versions of this product

Catalog No. AAH6217206



Specifications

Specifications

L-Aspartic acid 4-methyl ester hydrochloride
C5H10ClNO4
3628668
QRBMPUYOGOCYDJ-DFWYDOINSA-N
(2S)-2-amino-4-methoxy-4-oxobutanoic acid;hydrochloride
12832742
Solid
5g
16856-13-6
MFCD00038972
2s-2-amino-4-methoxy-4-oxobutanoic acid hydrochloride, 4-methyl l-aspartate hydrochloride, aspartic acid, 4-methylester, hydrochloride, h-asp ome-oh hcl, h-asp ome-oh.hcl, l-aspartic acid 4-methyl ester hcl, l-aspartic acid beta-methyl ester hydrochloride, l-aspartic acid, 4-methyl ester, hydrochloride, s-2-amino-4-methoxy-4-oxobutanoic acid hydrochloride, s-2-amino-succinic acid 4-methyl ester hydrochloride
COC(=O)CC(C(=O)O)N.Cl
183.588
183.59
97%

Chemical Identifiers

16856-13-6
183.588
QRBMPUYOGOCYDJ-DFWYDOINSA-N
12832742
COC(=O)CC(C(=O)O)N.Cl
C5H10ClNO4
MFCD00038972
2s-2-amino-4-methoxy-4-oxobutanoic acid hydrochloride, 4-methyl l-aspartate hydrochloride, aspartic acid, 4-methylester, hydrochloride, h-asp ome-oh hcl, h-asp ome-oh.hcl, l-aspartic acid 4-methyl ester hcl, l-aspartic acid beta-methyl ester hydrochloride, l-aspartic acid, 4-methyl ester, hydrochloride, s-2-amino-4-methoxy-4-oxobutanoic acid hydrochloride, s-2-amino-succinic acid 4-methyl ester hydrochloride
(2S)-2-amino-4-methoxy-4-oxobutanoic acid;hydrochloride
Safety and Handling

Safety and Handling

EINECSNumber : 240-880-5

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

Documents