missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Aspartic acid 4-cyclohexyl ester, 95%

$43.03 - $675.45

Chemical Identifiers

CAS 112259-66-2
Molecular Formula C10H17NO4
Molecular Weight (g/mol) 215.249
MDL Number MFCD00190762
InChI Key IJAPBBOAGRLKMH-QMMMGPOBSA-N
Synonym h-asp ochex-oh, h-asp ochx-oh, l-aspartic acid beta-cyclohexyl ester, 2s-2-amino-4-cyclohexyloxy-4-oxobutanoic acid, ambotzhaa5960, pubchem18997, l-aspartic acid b-cyclohexyl ester, h-asp ochex-oh hydrochloride salt
PubChem CID 14283862
IUPAC Name (2S)-2-amino-4-cyclohexyloxy-4-oxobutanoic acid
SMILES C1CCC(CC1)OC(=O)CC(C(=O)O)N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6370706 View Documents Alfa Aesar™
H6370706
5g Each for $43.03
 
AAH6370714 View Documents Alfa Aesar™
H6370714
25g Each for $171.18
 
AAH6370722 View Documents Alfa Aesar™
H6370722
100g Each for $675.45
 
Specifications

Chemical Identifiers

112259-66-2
215.249
IJAPBBOAGRLKMH-QMMMGPOBSA-N
14283862
C1CCC(CC1)OC(=O)CC(C(=O)O)N
C10H17NO4
MFCD00190762
h-asp ochex-oh, h-asp ochx-oh, l-aspartic acid beta-cyclohexyl ester, 2s-2-amino-4-cyclohexyloxy-4-oxobutanoic acid, ambotzhaa5960, pubchem18997, l-aspartic acid b-cyclohexyl ester, h-asp ochex-oh hydrochloride salt
(2S)-2-amino-4-cyclohexyloxy-4-oxobutanoic acid

Specifications

112259-66-2
MFCD00190762
IJAPBBOAGRLKMH-QMMMGPOBSA-N
(2S)-2-amino-4-cyclohexyloxy-4-oxobutanoic acid
14283862
Solid
5g
C10H17NO4
h-asp ochex-oh, h-asp ochx-oh, l-aspartic acid beta-cyclohexyl ester, 2s-2-amino-4-cyclohexyloxy-4-oxobutanoic acid, ambotzhaa5960, pubchem18997, l-aspartic acid b-cyclohexyl ester, h-asp ochex-oh hydrochloride salt
C1CCC(CC1)OC(=O)CC(C(=O)O)N
215.249
215.25
95%
L-Aspartic acid 4-cyclohexyl ester