missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Aspartic acid 4-cyclohexyl ester, 95%

Manufacturer:  Alfa Aesar™ H6370722

 View more versions of this product

Catalog No. AAH6370722



Specifications

Specifications

L-Aspartic acid 4-cyclohexyl ester
C10H17NO4
h-asp ochex-oh, h-asp ochx-oh, l-aspartic acid beta-cyclohexyl ester, 2s-2-amino-4-cyclohexyloxy-4-oxobutanoic acid, ambotzhaa5960, pubchem18997, l-aspartic acid b-cyclohexyl ester, h-asp ochex-oh hydrochloride salt
C1CCC(CC1)OC(=O)CC(C(=O)O)N
215.249
215.25
95%
112259-66-2
MFCD00190762
IJAPBBOAGRLKMH-QMMMGPOBSA-N
(2S)-2-amino-4-cyclohexyloxy-4-oxobutanoic acid
14283862
Solid
100g

Chemical Identifiers

112259-66-2
215.249
IJAPBBOAGRLKMH-QMMMGPOBSA-N
14283862
C1CCC(CC1)OC(=O)CC(C(=O)O)N
C10H17NO4
MFCD00190762
h-asp ochex-oh, h-asp ochx-oh, l-aspartic acid beta-cyclohexyl ester, 2s-2-amino-4-cyclohexyloxy-4-oxobutanoic acid, ambotzhaa5960, pubchem18997, l-aspartic acid b-cyclohexyl ester, h-asp ochex-oh hydrochloride salt
(2S)-2-amino-4-cyclohexyloxy-4-oxobutanoic acid
SDS Documents