missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Aspartic acid 4-benzyl ester 1-tert-butyl ester hydrochloride, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6593706
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
L-Aspartic acid 4-benzyl ester 1-tert-butyl ester hydrochloride | |
White | |
MFCD00190761 | |
s-4-benzyl 1-tert-butyl 2-aminosuccinate hydrochloride, h-asp obzl-otbu.hcl, 4-benzyl 1-tert-butyl 2s-2-aminobutanedioate hydrochloride, c15h21no4.hcl, h-asp obzl-otbu hcl, h-asp obzl-otbu hydrochloride, s-4-benzyl 1-tert-butyl 2-aminosuccinate hcl, l-aspartic acid beta-benzyl ester alfa-t-butyl ester hcl, 4-benzyl 1-tert-butyl s-2-aminosuccinate hydrochloride, 4-benzyl 1-tert-butyl l-aspartate-hydrogen chloride 1/1 | |
Cl.CC(C)(C)OC(=O)C(N)CC(=O)OCC1=CC=CC=C1 | |
315.79 | |
315.79 | |
5 g |
94347-11-2 | |
C15H22ClNO4 | |
2538326 | |
YSDLFDMXCRFHTQ-UHFFFAOYNA-N | |
4-benzyl 1-tert-butyl 2-aminobutanedioate hydrochloride | |
56777343 | |
98% | |
Solid |
Chemical Identifiers
94347-11-2 | |
315.79 | |
YSDLFDMXCRFHTQ-UHFFFAOYNA-N | |
56777343 | |
Cl.CC(C)(C)OC(=O)C(N)CC(=O)OCC1=CC=CC=C1 |
C15H22ClNO4 | |
MFCD00190761 | |
s-4-benzyl 1-tert-butyl 2-aminosuccinate hydrochloride, h-asp obzl-otbu.hcl, 4-benzyl 1-tert-butyl 2s-2-aminobutanedioate hydrochloride, c15h21no4.hcl, h-asp obzl-otbu hcl, h-asp obzl-otbu hydrochloride, s-4-benzyl 1-tert-butyl 2-aminosuccinate hcl, l-aspartic acid beta-benzyl ester alfa-t-butyl ester hcl, 4-benzyl 1-tert-butyl s-2-aminosuccinate hydrochloride, 4-benzyl 1-tert-butyl l-aspartate-hydrogen chloride 1/1 | |
4-benzyl 1-tert-butyl 2-aminobutanedioate hydrochloride |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only