Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Aspartic acid 1-benzyl ester, 98%

Chemical Identifiers

Chemical Name or Material L-Aspartic acid 1-benzyl ester
CAS 7362-93-8
Formula Weight 223.23
MDL Number MFCD00063186
Beilstein 2506965
Synonym H-Asp-OBzl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6312803

 
alfa aesar™
H6312803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6312806

 
alfa aesar™
H6312806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 223.23
Chemical Name or Material L-Aspartic acid 1-benzyl ester
Physical Form Solid
Assay 98%
MDL Number MFCD00063186
CAS 7362-93-8
Beilstein 2506965
Synonym H-Asp-OBzl
Quantity 1g
Storage Note 1 -30° to -10°C

Applications
L-Aspartic acid 1-benzyl ester, is used as an intermediate in organic synthesis. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in HCI solution.

Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Safety and Handling

-30° to -10°C