missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Aspartic acid 1-benzyl ester, 98%

Manufacturer:  Alfa Aesar™ H6312806

 View more versions of this product

Catalog No. AAH6312806



Description

Description

Applications
L-Aspartic acid 1-benzyl ester, is used as an intermediate in organic synthesis. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in HCI solution.

Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Specifications

L-Aspartic acid 1-benzyl ester
C11H13NO4
2506965
NJSRYBIBUXBNSW-VIFPVBQESA-N
(3S)-3-amino-4-oxo-4-phenylmethoxybutanoic acid
6994684
Solid
5g
7362-93-8
MFCD00063186
h-asp-obzl, l-aspartic acid benzyl ester, l-aspartic acid 1-benzyl ester, s-3-amino-4-benzyloxy-4-oxobutanoic acid, 1-benzyl l-aspartate, 3s-3-amino-4-oxo-4-phenylmethoxybutanoic acid, 3s-3-amino-4-benzyloxy-4-oxobutanoic acid, h-asp-obzl.hcl, h-asp obzl, pubchem14915
C1=CC=C(C=C1)COC(=O)C(CC(=O)O)N
223.228
223.23
98%

Chemical Identifiers

7362-93-8
223.228
NJSRYBIBUXBNSW-VIFPVBQESA-N
6994684
C1=CC=C(C=C1)COC(=O)C(CC(=O)O)N
C11H13NO4
MFCD00063186
h-asp-obzl, l-aspartic acid benzyl ester, l-aspartic acid 1-benzyl ester, s-3-amino-4-benzyloxy-4-oxobutanoic acid, 1-benzyl l-aspartate, 3s-3-amino-4-oxo-4-phenylmethoxybutanoic acid, 3s-3-amino-4-benzyloxy-4-oxobutanoic acid, h-asp-obzl.hcl, h-asp obzl, pubchem14915
(3S)-3-amino-4-oxo-4-phenylmethoxybutanoic acid
Safety and Handling

Safety and Handling

storageNote1 : -30° to -10°C

SDS Documents