Please login to your online account to display your discounted pricing

L-(+)-Asparagine hydrate, 98%, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material L-(+)-Asparagine hydrate
CAS 5794-13-8
Molecular Formula H2NCOCH2CH(NH2)CO2H·H2O
Formula Weight 150.14g/mol
MDL Number MFCD00151038
Merck Index 14,837
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA3651222

 
alfa aesar
36512-22
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 233° to235°C
Quantity 100g
Chemical Name or Material L-(+)-Asparagine hydrate
CAS 5794-13-8
Physical Form Powder
Assay 98%
Molecular Formula H2NCOCH2CH(NH2)CO2H·H2O
Formula Weight 150.14g/mol
MDL Number MFCD00151038
Merck Index 14,837
TSCA TSCA