Please login to your online account to display your discounted pricing

Alfa Aesar™ L-(+)-Asparagine hydrate, 98%

Chemical Identifiers

Chemical Name or Material L-(+)-Asparagine hydrate
CAS 5794-13-8
Formula Weight 150.14g/mol
MDL Number MFCD00151038
Merck Index 14,837
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3651214

 
alfa aesar™
36512-14
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3651236

 
alfa aesar™
36512-36
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 233° to 235°C
Quantity 25g
Chemical Name or Material L-(+)-Asparagine hydrate
CAS 5794-13-8
Physical Form Powder
Assay 98%
Formula Weight 150.14g/mol
MDL Number MFCD00151038
Merck Index 14,837
TSCA TSCA