Please login to your online account to display your discounted pricing

L-Asparagine, 99%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC371601000

 View more versions of this product

Catalog No. AC371601000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5% (on dry substance)
Physical Form Crystalline Powder
Formula Weight 132.12
Quantity 100g
Color White
Specific Rotation Condition + 34.50 (20.00°C c=10,2N HCl)
Packaging Glass bottle
Water 1% max.
Specific Rotation +33° to +36° (20°C, 589nm) (c=10, 2N HCl)
Assay Percent Range 99%
Chemical Name or Material L-Asparagine
CAS 70-47-3
MDL Number MFCD00064401
Infrared Spectrum Authentic
Linear Formula H2NCOCH2CH(NH2)CO2H
Molecular Formula C4H8N2O3

Specifications

Chemical Name or Material L-Asparagine
Assay ≥98.5% (on dry substance)
Assay Percent Range 99%
Molecular Formula C4H8N2O3
Linear Formula H2NCOCH2CH(NH2)CO2H
Formula Weight 132.12
MDL Number MFCD00064401