Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Asp-Asp-rhodamine Trifluoroacetate Salt

Manufacturer: alfa aesar™  J67000-EXD

 View all options for this product

Catalog No. AAJ67000EXD

 Please call Customer Service at 1-800-234-7437 or send an email to for assistance.

Description & Specifications


Color White to Off-white
Format Lyophilized Powder
Product Type Cell Lysis Detergent
Storage Requirements −30° to −10°C
Quantity 0.5mg
Molecular Formula C28H25N4O9·CF3CO2H
Formula Weight 788.56g/mol
Synonym (Asp)2-R110; Rhodamine 110, bis-(L-aspartic acid amide), trifluoroacetic acid salt
Sensitivity Light Sensitive