Please login to your online account to display your discounted pricing

L-(+)-Ascorbic Acid, Powder, BAKER ANALYZED™ Biochemical Reagent, J.T.Baker™


High quality biochemicals for research use

Quantity 500g
Packaging Poly Bottle

Manufacturer: avantor performance  B58107

 View more versions of this product

Catalog No. 02003149

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 176.12
Molecular Formula C6H8O6
Chemical Name or Material L-(+)-Ascorbic Acid
Purity Grade BAKER ANALYZED™ Biochemical Reagent
Physical Form Powder
Packaging Poly Bottle
Quantity 500g
Specific Gravity 1L = 1.65kg
CAS 50-81-7

Specifications

Chemical Name or Material L-(+)-Ascorbic Acid
CAS 50-81-7
Purity Grade BAKER ANALYZED™ Biochemical Reagent
Molecular Formula C6H8O6
Formula Weight 176.12