missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ L-(+)-Ascorbic acid, Electrophoresis Grade, ≥99%

Manufacturer:  Alfa Aesar™ J6629122

 View more versions of this product

Catalog No. AAJ6629122



Specifications

Specifications

L-(+)-Ascorbic acid
White
100g
C6H8O6
84272
ascoltin, ascorbate, ascorbic acid, ascorbicap, cevitamic acid, hybrin, l +-ascorbic acid, l-ascorbate, l-ascorbic acid, vitamin c
CIWBSHSKHKDKBQ-JLAZNSOCSA-N
(2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one
54670067
176.12
≥99%
1.65 g/mL
190°C to 193°C (decomposition)
50-81-7
MFCD00064328
14,830
Soluble in water at 0;5 M
C(C(C1C(=C(C(=O)O1)O)O)O)O
176.124
CHEBI:29073
Powder
Electrophoresis

Chemical Identifiers

50-81-7
176.124
CIWBSHSKHKDKBQ-JLAZNSOCSA-N
54670067
(2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one
C6H8O6
MFCD00064328
ascoltin, ascorbate, ascorbic acid, ascorbicap, cevitamic acid, hybrin, l +-ascorbic acid, l-ascorbate, l-ascorbic acid, vitamin c
CHEBI:29073
C(C(C1C(=C(C(=O)O1)O)O)O)O
Safety and Handling

Safety and Handling

EINECSNumber : 200-066-2

RTECSNumber : CI7650000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents