Please login to your online account to display your discounted pricing

L-(+)-Ascorbic acid, ACS, 99+%, Alfa Aesar™

$55.27 - $55.27

Chemical Identifiers

Chemical Name or Material L-(+)-Ascorbic acid
CAS 50-81-7
Purity Grade ACS Grade
Molecular Formula C6H8O6
Formula Weight 176.12
MDL Number MFCD00064328
Beilstein 84272
Merck Index 14,830
Synonym Vitamin C
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA3623722

 
alfa aesar
3623722
Each for $55.27
Description & Specifications

Specifications

Purity Grade ACS Grade
Formula Weight 176.12
Quantity 100g
Chemical Name or Material L-(+)-Ascorbic acid
TSCA TSCA
Density 1.65
Physical Form Powder
Assay ≥99%
Molecular Formula C6H8O6
MDL Number MFCD00064328
CAS 50-81-7
Synonym Vitamin C
Melting Point ≈191°C (dec.)
Beilstein 84272
Merck Index 14,830
RTECS Number CI7650000