missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Ascorbic acid, ≥98%, MP Biomedicals™
Manufacturer: Mp Biomedicals 0210076980
Specifications
L-Ascorbic acid | |
White to Yellow | |
50-81-7 | |
C6H8O6 | |
ascoltin, ascorbate, ascorbic acid, ascorbicap, cevitamic acid, hybrin, l +-ascorbic acid, l-ascorbate, l-ascorbic acid, vitamin c | |
C(C(C1C(=C(C(=O)O1)O)O)O)O | |
176.124 | |
CHEBI:29073 | |
Crystalline Powder |
1.65g/cm3 | |
100g | |
≥98% | |
MFCD00064328 | |
CIWBSHSKHKDKBQ-JLAZNSOCSA-N | |
(2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one | |
54670067 | |
176.1 |
Chemical Identifiers
50-81-7 | |
176.124 | |
CIWBSHSKHKDKBQ-JLAZNSOCSA-N | |
54670067 | |
(2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one |
C6H8O6 | |
MFCD00064328 | |
ascoltin, ascorbate, ascorbic acid, ascorbicap, cevitamic acid, hybrin, l +-ascorbic acid, l-ascorbate, l-ascorbic acid, vitamin c | |
CHEBI:29073 | |
C(C(C1C(=C(C(=O)O1)O)O)O)O |
Safety and Handling
EINECSNumber : 200-066-2
storageNote1 : Protect from light.
Recommended Storage : Store at room temperature.