missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-(+)-Ascorbic acid, 98+%, Alfa Aesar™

Manufacturer:  Alfa Aesar 1118822

 View more versions of this product

Catalog No. AA1118822



Specifications

Specifications

L-(+)-Ascorbic acid
~191°C (decomposition)
50-81-7
MFCD00064328
14,830
CIWBSHSKHKDKBQ-JLAZNSOCSA-N
(2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one
54670067
176.12
≥98%
1.65g/mL
100g
C6H8O6
84272
l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin
C(C(C1C(=C(C(=O)O1)O)O)O)O
176.124
CHEBI:29073
Powder

Chemical Identifiers

50-81-7
176.124
CIWBSHSKHKDKBQ-JLAZNSOCSA-N
54670067
(2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one
C6H8O6
MFCD00064328
l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin
CHEBI:29073
C(C(C1C(=C(C(=O)O1)O)O)O)O
Safety and Handling

Safety and Handling

RTECSNumber : CI7650000

TSCA : TSCA

Documents